Learn More
GSC Go Science Crazy Helmholtz Coil
GSC Go Science Crazy Helmholtz Coils are useful for creating near-uniform magnetic fields or for cancelling
Manufacturer:  GSC Go Science Crazy HEMCOIL
Catalog No. S39005
Add to cart
Description
Description
Features two 5-inch solenoids each made of 440 turns of enameled copper, a 26cm printed scale and two 4mm induction safety sockets