Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HELT Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548608

Catalog No. PA548608


Add to cart

Description

Description

Predicted to react with zebrafish based on sequence homology.

Transcriptional repressor which binds preferentially to the canonical E box sequence 5 ft.-CACGCG-3 ft..
Specifications

Specifications

HELT
Polyclonal
PBS with 0.09% sodium azide
Q7TS99
Hairy and enhancer of split-related protein HELT, HES/HEY-like transcription factor, HELT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
234219
Western Blot
Unconjugated
Helt
Liquid
HELT
KLH conjugated synthetic peptide between 131-160 amino acids from the Central region of human HELT.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit