Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hemacytometer Cover Glasses

Use alkali-free Thermo Scientific™ Hemacytometer Cover Glasses with red blood cell and platelet counting chambers.

Specifications

Thickness (English) 0.016 in.
Thickness (Metric) 0.4mm
View More Specs

Products 1
Catalog Number Mfr. No. Shape Price Quantity & Availability  
Catalog Number Mfr. No. Shape Price Quantity & Availability  
22-021801
View Documents
Thermo Scientific™
2026H001
Rectangular This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Use alkali-free Thermo Scientific™ Hemacytometer Cover Glasses with red blood cell and platelet counting chambers.

  • Precision cut
  • 0.016 in. (0.4mm) thickness
  • Two shapes available
Specifications

Specifications

0.016 in.
0.4mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit