Learn More
BioworldSupplier Diversity Partner Hemagglutinin-Influenza A Virus H7N7 Netherlands 219/03 (Recombinant), 2 μg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600911
Recombinant Hemagglutinin- Influenza A Virus H7N7 Netherlands (HA-Netherlands) Source: Baculovirus Insect Cells. Description : Recombinant HA- H7N7 Netherlands 219/03 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is 57,500 dalton. Physical Appearance: Sterile filtered colorless solution.Formulation: The protein solution contains 30mM Sodium phosphate, pH 7 and 500mM NaCl. Stability: Should be stored at 4°C.Purity: Greater than 90.0% as determined by SDS-PAGE.Immunological Activity: Western Blot 0.1 µg-1µg per strip, ELISA 1µg/well.Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-192
Due to product restrictions, please to purchase or view availability for this product.