Drummond Scientific Hemato-Clad™ Mylar™-Wrapped Hematocrit Tubes

Combine the precision of glass with the safety of Mylar wrapping

Manufacturer: Drummond Scientific 10007500HC/5

 View more versions of this product

Catalog No. 21-176-6

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance FDA, NIOSH, CDC, OSHA
    Color Red
    Diameter (English) Inner 0.04 to 0.047 in.
    Diameter (Metric) Inner 0.11 to 0.12cm
    For Use With (Application) For Blood collection
    Length (English) 2.95 in.
    Length (Metric) 7.5cm
    Material Glass
    Product Type Mylar™ Wrapped Hematocrit Tube, Heparin Coated

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit