Learn More
Hematopoietic Prostaglandin D Synthase/HPGDS Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF6487SP
Catalog No. AF6487SP
Add to cart
This item is not returnable. View return policy
Description
Description
Hematopoietic Prostaglandin D Synthase/HPGDS Polyclonal antibody AF6487SP specifically detects Hematopoietic Prostaglandin D Synthase/HPGDS in Human samples. It is validated for Western Blotting
Specifications
Specifications
Hematopoietic Prostaglandin D Synthase/HPGDS
LYOPH
Western Blot 1 ug/mL
O60760
EC 2.5.1.18, EC 5.3.99.2, Glutathione S-transferase, glutathione S-transferase sigma, Glutathione-dependent PGD synthase, Glutathione-requiring prostaglandin D synthase, GST class-sigma, GSTS, GSTShematopoietic prostaglandin D2 synthase, hematopoietic prostaglandin D synthase, H-PGDS, PGDS, PGDSglutathione-dependent PGD synthetase, Prostaglandin-H2 D-isomerase, PTGDS2
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Hematopoietic Prostaglandin D Synthase/HPGDS in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HPGDS
E. coli-derived recombinant human Hematopoietic Prostaglandin D Synthase/HPGDS Pro2-Leu199 Accession # O60760
Immunogen affinity purified
RUO
Primary
27306
Sterile PBS to a final concentration of 0.2 mg/mL.