Alfa Aesar™ Hematoxylin hydrate, 80% (dry wt.), water <8%

Manufacturer: Alfa Aesar™ 4722314

Quantity 25g

 View more versions of this product

Catalog No. AA4722314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 517-28-2
    Formula Weight 302.29 (Anhydrous)
    MDL Number MFCD00078111
    Beilstein 91399
    Merck Index 14,4637
    Synonym C.I. 75290; Natural Black 1
    Chemical Name or Material Hematoxylin hydrate, Water <8%

    Specifications

    Formula Weight 302.29 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00078111
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder
    CAS 517-28-2
    Beilstein 91399
    Merck Index 14,4637
    Synonym C.I. 75290; Natural Black 1
    Health Hazard 1 Warning
    Sensitivity Light Sensitive
    Color Cream to brown
    TSCA TSCA
    RTECS Number MH7875000
    Melting Point ≈200°C (dec.)
    Quantity 25g
    Chemical Name or Material Hematoxylin hydrate, Water <8%
    Assay 80%, (dry weight)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit