Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner HEME OXYGENASE INHIBITOR KIT  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner FHOITEAPK
The Heme Oxygenase Inhibitor Toolkit contains three inhibitors to heme oxygenase and a negative control [Cu (II) Protoporphyrin IX]. Heme Oxygenase catalyzes the conversion of heme to biliverdin in the heme degradation pathway and has been implicated in tumor cell resistance to chemotherapy, reduction of free radical formation and inflammation, and is associated with vesicular repair. Heme Oxygenase Inhibitor Toolbox contains: Zn (II) Protoporphyrin IX 100 mg, Cu (II) Protoporphyrin IX 50 mg, Sn (IV) Protoporphyrin IX Dichloride 250 mg, Zn (II) Deuteroepophyrin IX 2,4 bis ethylenglycol 25 mg
Catalog No. 50-720-345