Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Hemin, 99%, porcine

Catalog Number AC345960050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
16009-13-5
C34H32FeN4O4
616.50
MFCD00010726
GGIDWJQWCUJYRY-UHFFFAOYSA-L
chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
131675604
3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoic acid;iron;hydrochloride
[Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O
16009-13-5
C34H32FeN4O4
616.50
MFCD00010726
GGIDWJQWCUJYRY-UHFFFAOYSA-L
chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
131675604
3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoic acid;iron;hydrochloride
[Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O

Hemin, 98%, porcine, CAS # 16009-13-5, is an oxidized version of heme. It is an iron-containing prosthetic group in several proteins, and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hemin derived from porcine is an iron-containing prosthetic group present in some proteins and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion

Applications

• Hemin is a protoporphyrin IX containing a ferric iron (Fe3+) ion with a coordinating chloride ligand

• It is used in cellular protection and controls biological mechanisms

• It can stimulate globulin synthesis

• It can be applied to study bacterial strains and growth conditions in the short-term colony assays and cell transfections

• It is used during the preparation of agar medium for bacteria culture

• This compound has been used in L929 and J774 murine macrophage cells in order to study conjugation, characterization and toxicity of lipophosphoglycan-polyacrylic acid conjugate for vaccination against leishmaniasis

TRUSTED_SUSTAINABILITY
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 4679
Formula Weight 651.95
Percent Purity ≥97.5% (HPLC)
Physical Form Liquid
Chemical Name or Material Hemin, 99%
EINECSNumber 240-140-1
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit