Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemin, 99%, porcine, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
16009-13-5
C34H34ClFeN4O4-
653.965
MFCD00010726
WABFTRRMMIIYHI-UHFFFAOYSA-M
chlorohemin,hemin chloride,protoferriheme,ferriheme,ferriprotoporphyrin ix chloride,ferriprotoporphyrin ix,panhematin,protohemin,protohemin ix,ferriprotoporphyrin
131675604
3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoic acid;iron;hydrochloride
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)O.Cl.[Fe]
16009-13-5
C34H34ClFeN4O4-
653.965
MFCD00010726
WABFTRRMMIIYHI-UHFFFAOYSA-M
chlorohemin,hemin chloride,protoferriheme,ferriheme,ferriprotoporphyrin ix chloride,ferriprotoporphyrin ix,panhematin,protohemin,protohemin ix,ferriprotoporphyrin
131675604
3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoic acid;iron;hydrochloride
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)O.Cl.[Fe]
Quantity 25g
Infrared Spectrum Authentic
Color Colorless
Packaging Glass bottle
Assay Percent Range 98%
Merck Index 15, 4679
Formula Weight 651.95
Physical Form Liquid
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Hemin, 99%

EINECSNumber : 240-140-1

TSCA : TSCA