Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemin, Bovine, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
16009-13-5
C34H32FeN4O4
616.50
GGIDWJQWCUJYRY-UHFFFAOYSA-L
iron(3+) 4,20-bis(2-carboxyethyl)-9,14-diethenyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide
[Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O
16009-13-5
C34H32FeN4O4
616.50
GGIDWJQWCUJYRY-UHFFFAOYSA-L
iron(3+) 4,20-bis(2-carboxyethyl)-9,14-diethenyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide
[Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O

H1003, 16009-13-5, C34H32ClFeN4O4

Spectrum™ Hemin, Bovine , an iron-containing porphyrin, is used in the management of porphyria attacks. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 1