Learn More
Boditech Med hemochroma PLUS Microcuvettes
Microcuvettes for the hemochroma PLUS provides immediate access to hemoglobin and calculated hematocrit results.
Manufacturer:  Boditech Med 100202
Catalog No. 23-038-074
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Microcuvette shelf life of up to 2 years, even after the canister has been opened
Specifications
Specifications
100-101 Hemochroma PLUS Analyzer