Learn More
Boditech Med hemochroma PLUS Promo
The hemochroma PLUS provides immediate access to hemoglobin and calculated hematocrit results.
Manufacturer:  Boditech Med 100PROMO
Catalog No. 23-038-071
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Cleared for patients 6 months or older
  • Reagent-free microcuvettes
  • Microcuvette shelf life of up to 2 years, even after the canister has been opened
  • Measurement range 5.0 to 25.6 g/dL
  • 3.25 in. Color LCD Display
  • CLIA waived
Specifications
Specifications
24 month, 2 years
Small - 15 µL