Learn More
HemoCue™ Glucose 201 and WBC Analyzer Microcuvettes
Serves as a sample container and reaction chamber
Manufacturer:  HemoCue America 110705
 View more versions of this product
Catalog No. 22-601-005
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Serves as a sample container and reaction chamber
15° to 35°C
MicroCuvette
HemoCue Glucose 201 analyzer
Plastic
5μL