Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
HemoCue™ Plasma/Low Hb Photometer: Microcuvettes
For measurements of low concentrations of hemoglobin in plasma, serum or aqueous solutions
Manufacturer:  HemoCue America 110302
 View more versions of this product
Catalog No. 22-601-022
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Cuvettes Hb 201 analyzer
200
15 to 30°C
Plasma/low Hb analyzer
59 to 85°F
20μL