Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heneicosanoic acid, ≥99% (capillary GC), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco H51491G
Description
Food and BeveragesSpecifications
Poly Bottle | |
2363-71-5 | |
CH3(CH2)19 COOH | |
326.56 | |
Analytical Standard | |
Ambient |
1 g | |
Analytical Standard | |
MFCD00002805 | |
≥99% (capillary GC) | |
74°C to 75°C | |
Neat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction