Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heneicosanoic acid, ≥99% (capillary GC), MilliporeSigma™ Supelco™

$40.65 - $224.00

Specifications

Percent Purity ≥99% (capillary GC)
Grade Analytical Standard
Linear Formula CH3(CH2)19 COOH
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3635
View Documents
MilliporeSigma Supelco
H5149100MG
100 mg
Each for $40.65
Add to cart
 
11-101-3634
View Documents
MilliporeSigma Supelco
H51491G
1 g
Each for $224.00
Add to cart
 
Description

Description

Food and Beverages
Specifications

Specifications

≥99% (capillary GC)
Analytical Standard
CH3(CH2)19 COOH
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit