Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HENMT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555866
Description
Immunogen sequence: MEENNLQCSS VVDGNFEEVP RETAIQFKPP LYRQRYQFVK NLVDQHEPKK VADLGCGDTS LLRLLKVNPC IELLVGV Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 47%.

Specifications
HENMT1 | |
Polyclonal | |
Unconjugated | |
HENMT1 | |
4921515J06Rik; C1orf59; HEN1; HEN1 methyltransferase homolog 1; HEN1 methyltransferase homolog 1 (Arabidopsis); Henmt1; hua enhancer 1 homolog 1; mHEN1; RGD1306230; Small RNA 2'-O-methyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T8I9 | |
HENMT1 | |
Recombinant protein corresponding to Human HENMT1. Recombinant protein control fragment (Product #RP-94398). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction