Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ HEPA Air Filter
SDP

Fresh air supply for unit; filter classification H14.

$2157.50

Specifications

Weight (English) 2.20 lb.
Weight (Metric) 1 kg
Description HEPA Air Filter
View More Specs

Products 4
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
01-672-903
SDP
View Documents
BINDER™
80122314
BINDER Units M 720
Each for $2,157.50
Add to Cart
 
01-672-904
SDP
View Documents
BINDER™
80122313
BINDER Units M 260
Each for $2,157.50
Add to Cart
 
01-672-905
SDP
View Documents
BINDER™
80122312
BINDER Units M 115
Each for $2,157.50
Add to Cart
 
01-672-906
SDP
View Documents
BINDER™
80122311
BINDER Units M 56
Each for $2,157.50
Add to Cart
 
Description

Description

  • HEPA Air Filter
  • For use with various BINDER™ Unit M lines
Specifications

Specifications

2.20 lb.
1 kg
HEPA Air Filter
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.