Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits Available on GSA/VA Contract for Federal Government customers only.
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
Manufacturer: Thermo Scientific™ 760209
 View more versions of this product
Catalog No. 15-497-023
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With 8000 Series models
    Description Main HEPA Filter, Value pack (4/pk)
    Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
    HEPA Filters
    • 99.97% filtering at 0.3µm
    • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
    • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
    Decontamination Kits
    • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
    • Reduces downtime and labor spent on decontamination
  • Description & Specifications
Related Products
Product Content Feedback