Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparan Sulfate Proteoglycan 2 Mouse anti-Human, Bovine, Alexa Fluor™ 750, Clone: A74, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223523AF750

Catalog No. NP22323A70


Add to cart

Description

Description

Heparan Sulfate Proteoglycan 2 Monoclonal antibody specifically detects Heparan Sulfate Proteoglycan 2 in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Heparan Sulfate Proteoglycan 2
A74
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Conjugated, Purified
HSPG2
Bovine corneal endothelial cells
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Clone A74 is highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). Clone A74 cross-reacts with human perlecan. Other species have not been tested.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1
Mouse
IgG1
0.1 mL
Primary
3339.0
Bovine, Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit