Learn More
HeparanSulfateProteoglycan2 Mouse anti-Bovine, Janelia Fluor 549, Clone: A74, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223523JF549
Catalog No. NBP23523549
Add to cart
Description
Description
HeparanSulfateProteoglycan2 Monoclonal antibody specifically detects HeparanSulfateProteoglycan2 in Bovine samples. It is validated for Immunohistochemistry.
Specifications
Specifications
HeparanSulfateProteoglycan2
A74
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
HSPG2
Bovine corneal endothelial cells
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Clone A74 is highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). Clone A74 cross-reacts with human perlecan. Other species have not been tested.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1
Mouse
IgG1
0.1mL
Primary
3339
Bovine, Human