Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heparin Sodium Salt, Thermo Scientific™

Click to view available options
Quantity:
1mU
50kU
100kU
500kU
Packaging:
Plastic bottle
9042-11-9
MFCD00081689
9042-11-9
MFCD00081689
Physical Form Powder
Packaging Plastic bottle
Quantity 50kU
Chemical Name or Material Heparin Sodium Salt