Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis A Surface Antigen Antibody (BGN/B5B3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062197JF646
Description
Hepatitis A Surface Antigen Monoclonal antibody specifically detects Hepatitis A Surface Antigen in Virus samples. It is validated for ELISASpecifications
Hepatitis A Surface Antigen | |
Monoclonal | |
Janelia Fluor 646 | |
Mouse | |
Protein G purified | |
RUO | |
Virus | |
Purified |
ELISA | |
BGN/B5B3 | |
50mM Sodium Borate | |
Lysate from foetal kidney cells of rhesus monkey infected with extract from patient with active viral Hepatitis A. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction