Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Antigen Mouse anti-Virus, Clone: 4H9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP179176

 View more versions of this product

Catalog No. NBP179176


Add to cart

Description

Description

Hepatitis B Virus X antigen Monoclonal antibody specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot.
Specifications

Specifications

Hepatitis B Virus X antigen
4H9
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
HBVgp3
The specific Immunogen is proprietary information.
17 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 200ul DI water. 20ul sample is not lyoph and is a liquid.
ELISA, Western Blot
LYOPH
Western Blot 1:1000
NP_647606
HBVgp3 X protein, HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein
Mouse
IgG
Protein A or G purified
RUO
Primary
944566
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit