Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Antigen Mouse anti-Virus, Clone: 4H9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP17917620UL

 View more versions of this product

Catalog No. NBP17917620


Add to cart

Description

Description

Hepatitis B Antigen Monoclonal antibody specifically detects Antigen in Virus samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

Hepatitis B Antigen
4H9
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
Mouse
IgG
Protein A or G purified
RUO
Primary
Reconstitute with 200ul DI water. 20ul sample is not lyoph and is a liquid.
ELISA, Western Blot
LYOPH
Western Blot 1:1000, ELISA 1:2000
NP_647606
HBV X antigen, HBx, HBxAg
The specific Immunogen is proprietary information.
17 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit