Learn More
Hepatitis B envelope antigen Mouse anti-Hepatitis B virus, Clone: HM028, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13565
Catalog No. 89-033-528
Add to cart
Description
Description
Mouse monoclonal antibody raised against hepatitis B envelope antigen (HBeAg).
  • High specificity
  • High reactivity
Specifications
Specifications
Hepatitis B envelope antigen
HM028
Mouse monoclonal antibody raised against hepatitis B envelope antigen (HBeAg).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with HBeAg.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG1
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Hepatitis B virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)