Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B surface antigen Goat anti-Hepatitis B virus, Polyclonal, Abnova™

Goat Polyclonal Antibody

$346.00 - $469.00

Specifications

Antigen Hepatitis B surface antigen
Host Species Goat
Target Species Hepatitis B virus
Applications ELISA, Immunofluorescence, Western Blot
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-033-694 View Documents Abnova Corporation
PAB29849
1 mg Each for $469.00
Add to cart
 
89-033-695 View Documents Abnova Corporation
PAB29850
500 uL Each for $346.00
Add to cart
 
Description

Description

Goat polyclonal antibody raised against hepatitis B surface antigen (HBsAg).

  • High specificity
  • High reactivity
Specifications

Specifications

Hepatitis B surface antigen
Hepatitis B virus
RUO
Liquid
Primary
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Goat
ELISA, Immunofluorescence, Western Blot
Unconjugated
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit