Learn More
Hepatitis B surface antigen Mouse anti-Hepatitis B virus, Clone: FK314, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13559
Catalog No. 89-033-522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mouse monoclonal antibody raised against hepatitis B surface antigen (HBsAg).
  • High specificity
  • High reactivity
Specifications
Specifications
Hepatitis B surface antigen
Unconjugated
In 10 mM PBS, 50 mM NaCl, pH 8.0 (0.05% sodium azide).
Mouse
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Hepatitis B virus
FK314
Mouse monoclonal antibody raised against hepatitis B surface antigen (HBsAg).
Liquid
IgA
RUO
Primary
This antibody reacts with HBsAg.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)