Learn More
Hepatitis B surface antigen Mouse anti-Hepatitis B virus, Clone: J343, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13562
Catalog No. 89-033-525
Add to cart
Description
Description
Mouse monoclonal antibody raised against hepatitis B surface antigen (HBsAg).
  • High specificity
  • High reactivity
Specifications
Specifications
Hepatitis B surface antigen
J343
Mouse monoclonal antibody raised against hepatitis B surface antigen (HBsAg).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
IgG2a
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Hepatitis B virus
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Native purified hepatitis B surface antigen (HBsAg).
Protein A purification
RUO
Primary
This antibody reacts with HBsAg of hepatitis B virus subtypes Ad and Ay.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)