Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B surface antigen Mouse anti-Hepatitis B virus, Clone: J345, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13564
Description
Mouse monoclonal antibody raised against hepatitis B surface antigen (HBsAg).
- High specificity
- High reactivity
Specifications
Hepatitis B surface antigen | |
Monoclonal | |
Unconjugated | |
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
Mouse | |
Protein A purification | |
RUO | |
This antibody reacts with HBsAg of hepatitis B virus subtypes Ad and Ay. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
ELISA, Immunofluorescence, Lateral Flow | |
J345 | |
Mouse monoclonal antibody raised against hepatitis B surface antigen (HBsAg). | |
In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
Native purified hepatitis B surface antigen (HBsAg). | |
1 mg | |
Primary | |
Hepatitis B virus | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction