Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Hepatitis B Virus Core Antigen (HBVcAg) Ab-1, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Hepatitis B Virus Core Antigen (HBVcAg) Ab-1, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB1413A1

 View more versions of this product

Catalog No. RB1413A1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Hepatitis B Virus Core Antigen (HBVcAg) Ab-1, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Hepatitis B virus is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Hepatitis B virus

Positive Control: Hepatitis B virus infected liver

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Hepatitis B Virus Core Antigen (HBVcAg) Ab-1
Polyclonal
Unconjugated
Rabbit
IgG
ASR
Primary
Polyclonal
Immunohistochemistry (Paraffin)
Polyclonal
Concentrated
Hepatitis B virus
500μL
Infectious Disease
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit