Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus X antigen Mouse anti-Virus, Alexa Fluor™ 594, Clone: X36C, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222445AF594

Catalog No. NP22244A94


Add to cart

Description

Description

Hepatitis B Virus X antigen Monoclonal antibody specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Hepatitis B Virus X antigen
X36C
Western Blot, ELISA
Affinity Purified, Conjugated
HBVgp3
Baculovirus expressed recombinant HBx.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Virus
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
HBVgp3 X protein, HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein
Mouse
IgG1
0.1 mL
Primary
944566.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit