Learn More
Invitrogen™ Hepatitis B Virus X Monoclonal Antibody (1884)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547492
Description
Specificity: Specific to HBx. Non-reactive with human cells. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The Hepatitis B X antigen is a 154 amino acid ~17 kDa multifunctional protein involved in the development of liver chirrosis and hepatocellular carcinoma. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
Specifications
Hepatitis B Virus X | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Western Blot, Immunocytochemistry | |
1884 | |
Unconjugated | |
HBV; HBVgp3; HepB; HepB X | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.