Learn More
BioworldSupplier Diversity Partner HEPATITIS C VIRUS NS3 GENOT DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220601501
HEPATITIS C VIRUS NS3 GENOTYPE-5A, (RECOMBINANT): Recombinant Hepatitis C Virus NS3 Genotype-5a Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNA virus in the family Flaviviridae. HCV has a ...
Catalog No. 50-253-251
Due to product restrictions, please to purchase or view availability for this product.