Learn More
BioworldSupplier Diversity Partner HEPATITIS C VIRUS NUCLEOCAP DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220601651
HEPATITIS C VIRUS NUCLEOCAPSID (CORE) 24, (RECOMBINANT): Recombinant Hepatitis C Virus Nucleocapsid 24 Source:E.coli Background: The hepatitis C virus (HCV) core protein represents the first 191 amino acids of the viral precursor pol...
Catalog No. 50-253-266
Due to product restrictions, please to purchase or view availability for this product.