Learn More
BioworldSupplier Diversity Partner HEPATITIS C VIRUS NUCLEOCAP DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220601371
HEPATITIS C VIRUS NUCLEOCAPSID (CORE) GENOTYPE-2A, (RECOMBINANT): Recombinant Hepatitis C Virus Nucleocapsid Genotype 2a (2-119 a.a) Source:E.coli Background: The hepatitis C virus (HCV) core protein represents the first 191 amino ac...
Catalog No. 50-253-238
Due to product restrictions, please to purchase or view availability for this product.