Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis E Virus Capsid Mouse anti-Virus, Clone: BD6.A2.A9.E11.F8, Novus Biologicals™

Mouse Monoclonal Antibody
$515.50
Specifications
Antigen | Hepatitis E Virus Capsid |
---|---|
Clone | BD6.A2.A9.E11.F8 |
Concentration | 1 mg/mL |
Dilution | ELISA |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB159187
![]() |
Novus Biologicals
NBP3148560.1MG |
0.1 mg |
Each for $515.50
|
|
|||||
NB159188
![]() |
Novus Biologicals
NBP3148560.5MG |
0.5 mg |
Each for $1,262.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Hepatitis E Virus Capsid Monoclonal antibody specifically detects Hepatitis E Virus Capsid in Virus samples. It is validated for ELISASpecifications
Hepatitis E Virus Capsid | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Virus | |
ORF2, pORF2, Secreted protein ORF2 | |
IgG1 κ | |
Protein G purified | |
Antibody |
BD6.A2.A9.E11.F8 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline, pH 7.4, Filter Sterile | |
HEV ORF2 aa1-660 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title