Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis G Virus Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093584JF549
Description
Hepatitis G Virus Polyclonal antibody specifically detects Hepatitis G Virus in Human, Virus samples. It is validated for Western BlotSpecifications
Hepatitis G Virus | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Human, Virus | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
Hepatitis GB virus C, HGV | |
Reacts with residues 2268-2276 [CDKCEARQE] of HGV. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction