Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis G Virus Rabbit anti-Human, Virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | Hepatitis G Virus |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10093584
|
Novus Biologicals
NB10093584 |
0.1 mg |
Each for $378.00
|
|
NB017932
|
Novus Biologicals
NB10093584SS |
0.025 mg |
Each for $177.16
|
|
Description
Hepatitis G Virus Polyclonal specifically detects Hepatitis G Virus in Human, Virus samples. It is validated for Western Blot.Specifications
Hepatitis G Virus | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Human, Virus | |
Hepatitis GB virus C, HGV | |
IgG | |
Affinity Purified | |
Hepatitis G Virus |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Reacts with residues 2268-2276 [CDKCEARQE] of HGV. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title