Learn More
Bioworld HEPATITISA VIRUSVP3 (RECOMB
Supplier:  Bioworld 220601761
Encompass_Preferred
HEPATITISA VIRUSVP3 (RECOMBINANT): Recombinant Hepatitis A Virus VP3 (304-415 a.a) Source: E.Coli Background: HAV, the prototype of the genus Hepatovirus, belongs to the family Picornaviridae. Its 7.5-kb single-stranded RNA genome bea...
Catalog No. 50-253-276
Due to product restrictions, please to purchase or view availability for this product.