Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (EP-2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142179
Description
Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Hepatocyte Specific Antigen | |
Monoclonal | |
0.1 mg/mL | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Mouse | |
Protein A or G purified | |
RUO | |
Monoclonal antibody EP-2 stains human hepatocytes and the majority of human hepato-carcinomas. In frozen sections, it stains the cytoplasm of hepatic cells and may be used as a marker of the liver. Cell preparations of hepatocellular carcinoma biopsies or cell lines are found to bind EP-2 on the cell surface. Monoclonal antibodies to liver cell processes are useful in the identification of hepatic carcinomas and normal organ structures. Two monoclonal antibodies to different liver substructures are offered which bind to subsets of hepatic carcinomas. | |
Store at 4C. Do not freeze. | |
IgG2b |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
EP-2 | |
Unconjugated | |
Hepatocyte, HSA | |
The antibody reacts with the Hepatocytes antigen. | |
0.1 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction