Learn More
Hepatocyte Specific Antigen Mouse anti-Human, Alexa Fluor™ 750, Clone: HSA98, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234645AF750
Catalog No. NP23464A70
Add to cart
Description
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
Hepatocyte Specific Antigen
HSA98
Immunocytochemistry/Immunofluorescence
Hepatocyte, HSA
HEP-3B human hepatocellular carcinoma cells
0.1 mL
Primary
Monoclonal antibodies (MAbs) to liver cell processes are useful in the identification of hepatic carcinomas and normal organ structures. MAb HSA98 binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCCs). In frozen sections, it stains hepatic cells and may be used as a marker of the liver. Cell preparations of hepatocellular carcinoma biopsies or cell lines are found to bind HSA98 on the cell surface. This MAb stains liver hepatocytes in frozen human liver sections and is positive on the cell surface of human liver carcinomas.
Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human