Learn More
Hepatocyte Specific Antigen Mouse anti-Human, Canine, Alexa Fluor™ 750, Clone: OCH1E5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234699AF750
Catalog No. NP234699A70
Add to cart
Description
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human,Canine samples. It is validated for Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
Hepatocyte Specific Antigen
OCH1E5
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Hepatocyte, HSA
Extract of a formalin-fixed, rejected-allograft of a human liver
0.1 mL
Primary
Human, Canine
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal