Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HEPES, ≥99% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Plastic bottle
7365-45-9
C8H18N2O4S
238.302
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid]
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCO)CCS(=O)(=O)O
7365-45-9
C8H18N2O4S
238.302
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid]
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCO)CCS(=O)(=O)O
Beilstein 883043
Formula Weight 238.3
Physical Form Crystalline Powder
Percent Purity ≥99%
Packaging Plastic bottle
Melting Point ~236°C (decomposition)
Quantity 100g
Chemical Name or Material HEPES
Assay Dry basis

P261-P271-P304+P340-P312-P501c

H335

EINECSNumber : 230-907-9

RTECSNumber : TL6809000

TSCA : Yes

Recommended Storage : Ambient temperatures