Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ HEPES Buffer, 0.2M buffer soln., pH 6.5, low endotoxin

Click to view available options
Quantity:
100mL
250mL
7365-45-9
C8H18N2O4S
238.302
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCO)CCS(=O)(=O)O
7365-45-9
C8H18N2O4S
238.302
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCO)CCS(=O)(=O)O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 6.5
Color Colorless
Quantity 250mL
Chemical Name or Material HEPES Buffer, low endotoxin

TSCA : TSCA

storageNote1 : 2° to 8°C