Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES Buffer, 0.2M buffer soln., pH 6.5, low endotoxin, Thermo Scientific™

Catalog Number AAJ67320AE
Click to view available options
Quantity:
100 mL
250 mL
7365-45-9
C8H18N2O4S
238.30
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
OCCN1CCN(CCS(O)(=O)=O)CC1
7365-45-9
C8H18N2O4S
238.30
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
OCCN1CCN(CCS(O)(=O)=O)CC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.2 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material HEPES Buffer, low endotoxin
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit