HEPES, Free Acid, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer: J.T.Baker 401800

Packaging Poly Bottle
Quantity 25g

 View more versions of this product

Catalog No. 02-002-970

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7365-45-9
    Molecular Formula C8H18N2O4S
    Formula Weight 238.31
    Chemical Name or Material HEPES, Free Acid, Free Acid
    Grade Ultrapure Bioreagent

    Specifications

    Formula Weight 238.31
    Molecular Formula C8H18N2O4S
    Quantity 25g
    CAS 7365-45-9
    Packaging Poly Bottle
    Chemical Name or Material HEPES, Free Acid, Free Acid
    Grade Ultrapure Bioreagent

    Description

      For Liquid Chromatography and Molecular Biology Applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit