Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, Free Acid, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle

Chemical Identifiers

7365-45-9
C8H18N2O4S
238.302
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology Applications
CAS 7365-45-9
Molecular Formula C8H18N2O4S
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES C1CN(CCN1CCO)CCS(=O)(=O)O
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Molecular Weight (g/mol) 238.302
PubChem CID 23831
ChEBI CHEBI:42334
Formula Weight 238.31
Packaging Poly Bottle
Quantity 100g
Chemical Name or Material HEPES, Free Acid, Free Acid
Grade Ultrapure Bioreagent
Show More Show Less