Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HEPES Lysis Buffer with Triton™ X-100, 2X

$73.78

Specifications

Concentration 2X
Format Liquid
Description Clear
Product Type Cell Lysis Reagent
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ62809EQE
View Documents
Thermo Scientific™
J62809EQE
125mL
Each for $73.78
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ62809AK
View Documents
Thermo Scientific™
J62809AK
250mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2X
Liquid
Clear
Cell Lysis Reagent
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit